Learn More
INTACT GENOMICS INC BL21(DE3) Electrocompetent Cells 12x100µl
Catalog No. 5000082875
Encompass
Intact Genomics BL21 (DE3) electrocompetent E. coli cells are suitable for high efficiency transformation and routine protein expression. Increased cloning efficiencies versus typical BL21 cells makes the BL21(DE3) electrocompetent E. coli cells ideal for construction of complex expression libraries.BL21(DE3) Electrocompetent Cells have the following features: >1 x 10^10 cfu/µg efficiency with electroporation Widely used host background T7 Expression Strain Deficient in both lon (1) and ompT proteases Resistant to phage T1 (fhuA2) B StrainProduct Includes and Storage:BL21(DE3) Electrocompetent Cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC
Catalog No. 50-000-82875 Supplier INTACT GENOMICS INC Supplier No. 125448
Intact Genomics BL21 (DE3) electrocompetent E. coli cells are suitable for high efficiency transformation and routine protein expression. Increased cloning efficiencies versus typical BL21 cells makes the BL21(DE3) electrocompetent E. coli cells ideal for construction of complex expression libraries.BL21(DE3) Electrocompetent Cells have the following features: >1 x 10^10 cfu/µg efficiency with electroporation Widely used host background T7 Expression Strain Deficient in both lon (1) and ompT proteases Resistant to phage T1 (fhuA2) B StrainProduct Includes and Storage:BL21(DE3) Electrocompetent Cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC