Learn More
INTACT GENOMICS INC BL21(DE3)pLysS ElectroCompetent Cells 6x100μl
Catalog No. 5000082899
Encompass
Intact Genomics (ig) BL21(DE3)pLysS Electrocompetent Cells are suitable for transformation and routine protein expression. The pLysS plasmid produces T7 lysozyme that reduces base level expression of the gene of interest, subsequently allowing for tighter control of expression and is therefore suitable for expression of toxic genes. BL21(DE3)pLysS Electrocompetent Cells also carry the chloramphenicol resistance gene.ig BL21(DE3)pLysS Electrocompetent Cells have the following features: T7 Expression Strain Deficient in both lon (1) and ompT proteases Resistant to phage T1 (fhuA2) B Strain Suitable for expression of toxic genesProduct Includes and Storage:ig BL21(DE3)pLysS Electrocompetent Cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC
Catalog No. 50-000-82899 Supplier INTACT GENOMICS INC Supplier No. 125624
Intact Genomics (ig) BL21(DE3)pLysS Electrocompetent Cells are suitable for transformation and routine protein expression. The pLysS plasmid produces T7 lysozyme that reduces base level expression of the gene of interest, subsequently allowing for tighter control of expression and is therefore suitable for expression of toxic genes. BL21(DE3)pLysS Electrocompetent Cells also carry the chloramphenicol resistance gene.ig BL21(DE3)pLysS Electrocompetent Cells have the following features: T7 Expression Strain Deficient in both lon (1) and ompT proteases Resistant to phage T1 (fhuA2) B Strain Suitable for expression of toxic genesProduct Includes and Storage:ig BL21(DE3)pLysS Electrocompetent Cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC