Learn More
ATCC BL3.1; B-Lymphosarcoma; Bovine (Bos taurus)
Catalog No. 502384509
Encompass
BL3.1 is a B lymphocyte cell that was isolated from a a 3-month-old male Hereford calf with lymphosarcoma. The cells can be used in BLV and retroviral studies and as a target for NK assays.
Catalog No. 50-238-4509 Supplier ATCC Supplier No. CRL2306
BL3.1 is a B lymphocyte cell that was isolated from a a 3-month-old male Hereford calf with lymphosarcoma. The cells can be used in BLV and retroviral studies and as a target for NK assays.