Learn More
Arcticwhite LLC 24 Well Assay Plate, Polystyrene, Clear Bottom, Black, 24 Wells, ANSI/SLAS Standard Base and Footprint, Black Opaque Wells, 68 Assay Plates per Case
SDP
Catalog No. NC0346907
Encompass
24 Well Assay Plate, Polystyrene, Clear Bottom, Black, 24 Wells, ANSI/SLAS Standard Base and Footprint, Black Opaque Wells, 68 Assay Plates per Case. This assay microplate is for optical and luminescent assays. Format - 24 wells, ANSI/SLAS standard base and footprint. Bottom - Clear. Material - Polystyrene. Color - Black opaque wells. Supplied by Arctic White as 68 assay plates per case.
Catalog No. NC0346907 Supplier Arcticwhite LLC Supplier No. AWLS303008
24 Well Assay Plate, Polystyrene, Clear Bottom, Black, 24 Wells, ANSI/SLAS Standard Base and Footprint, Black Opaque Wells, 68 Assay Plates per Case. This assay microplate is for optical and luminescent assays. Format - 24 wells, ANSI/SLAS standard base and footprint. Bottom - Clear. Material - Polystyrene. Color - Black opaque wells. Supplied by Arctic White as 68 assay plates per case.