Learn More
Research Products International Corp Opaque Black Acrylic Storage Box
SDP
Catalog No. 50998433
Encompass
Convenient storage box for organizing and protecting valuable antibodies and other laboratory samples that are light sensitive. Manufactured from opaque black light proof acrylic that is temperature resistant down to -70°C. Features a durable hinged lid with overhang for added protection from light.
Catalog No. 50-998-433 Supplier Research Products International Corp Supplier No. 145782
Convenient storage box for organizing and protecting valuable antibodies and other laboratory samples that are light sensitive. Manufactured from opaque black light proof acrylic that is temperature resistant down to -70°C. Features a durable hinged lid with overhang for added protection from light.