Learn More
Waters Corp Crimp Cap, Black, Aluminum, PTFE/Silicone Septum, 100/pk
Catalog No. 502392369 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a black, aluminum crimp cap with a PTFE/Silicone septum, supplied in a package of 100 units. It is designed for sample preparation applications and falls under the category of caps.
  • Crimp closure type
  • Black cap color
  • PTFE/Silicone seal material
  • Includes cap & septum
  • Designed for sample preparation
  • Supplied in a package of 100 units
Catalog No. 50-239-2369 Supplier Waters Corp Supplier No. 186006967
This product is a black, aluminum crimp cap with a PTFE/Silicone septum, supplied in a package of 100 units. It is designed for sample preparation applications and falls under the category of caps.
  • Crimp closure type
  • Black cap color
  • PTFE/Silicone seal material
  • Includes cap & septum
  • Designed for sample preparation
  • Supplied in a package of 100 units