Learn More
THORLABS INC Matte Black Aluminum Foil, 1' x 50' (305 mm x 15.2 m) x .002" (50 µm) Thick
Catalog No. NC0408932
Encompass
Matte Black Aluminum Foil, 1' x 50' (305 mm x 15.2 m) x .002" (50 µm) Thick
Catalog No. NC0408932 Supplier THORLABS INC Supplier No. BKF12
Matte Black Aluminum Foil, 1' x 50' (305 mm x 15.2 m) x .002" (50 µm) Thick