Learn More
KETCHUM MFG CO INC BLACK ANIMAL TATTOO PASTE-5 OZ
Catalog No. NC1933391
Encompass
Intended for tattooing of animals to create a black tattoo identification. Clean and dry area to be marked. Apply paste, use tattoo instrument of your choice, then rub more paste in tattoo punctures with a gloved finger or brush to insure a lasting tattoo.
Catalog No. NC1933391 Supplier KETCHUM MFG CO INC Supplier No. 327
Intended for tattooing of animals to create a black tattoo identification. Clean and dry area to be marked. Apply paste, use tattoo instrument of your choice, then rub more paste in tattoo punctures with a gloved finger or brush to insure a lasting tattoo.