Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Conductive Automation Tips for Qiagen™ Workstations
GREENER_CHOICE

Catalog No. 21402352 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
300 μL
Filtered:
Non-filtered
Fit for Purpose:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Fit for Purpose
21-402-352 300 μL Non-filtered Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-402-352 Supplier Thermo Scientific™ Supplier No. 951251
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.

  • Available in nonsterile, sterile and with the ART™ filter technology
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 300 μL
Filtered Non-filtered
Sterility Sterile
Color Black
Format Rack
Tip Style Conductive
Packaging 96 Tips/Tray, 10 Trays/Case
For Use With (Equipment) Universal Fit, Qiagen
Product Line Qiagen
Fit for Purpose Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.