Learn More
Psl Equipment Inc BLACK BIOPSY PAPER 500/PK
Catalog No. NC0280794
Encompass
Non-absorbent porous mesh, specifically designed for processing biopsy specimens. Allows fluid to flow through freely while containing small tissue fragments.
Catalog No. NC0280794 Supplier Psl Equipment Inc Supplier No. BPB
Non-absorbent porous mesh, specifically designed for processing biopsy specimens. Allows fluid to flow through freely while containing small tissue fragments.