Learn More
Alkali Scientific Western Blot Boxes, 3 1/2 X 2 9/16 X 1", Min Volume 4mL, Black, 5/pk
SDP
Catalog No. NC0887948
Encompass_Preferred
Western blot boxes and containers are opaque black for light sensitive samples. They fit nearly any sized blot and feature a hinged lid that easy to open and close. The corners are 90˚ and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot boxes. When you match the size of the box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.
Catalog No. NC0887948 Supplier Alkali Scientific Supplier No. BC12007BK
Western blot boxes and containers are opaque black for light sensitive samples. They fit nearly any sized blot and feature a hinged lid that easy to open and close. The corners are 90˚ and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot boxes. When you match the size of the box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.