Learn More
Uline Colored Bottle Brush | Black | 1 EA
Catalog No. NC2943654
Encompass
Strong polypropylene handle with extra-stiff polyester bristles, For dairy plants and bottling facilities, FDA compliant, Chemically resistant to acids, alkalis, alcohols, benzene, gasoline and most cleaning solvents, Aids in HACCP compliance
Catalog No. NC2943654 Supplier Uline Supplier No. H8558BL
Strong polypropylene handle with extra-stiff polyester bristles, For dairy plants and bottling facilities, FDA compliant, Chemically resistant to acids, alkalis, alcohols, benzene, gasoline and most cleaning solvents, Aids in HACCP compliance