Learn More
Perkin Elmer US LLC Black Bristle Brush - 9 mm
Catalog No. 502113250
Encompass_Preferred
Applications include: general dustings, cleaning, marking. This brush is suitable for use with IPA, trichloroethane and other cleaning solvents.
Catalog No. 50-211-3250 Supplier Perkin Elmer US LLC Supplier No. L9001026
Applications include: general dustings, cleaning, marking. This brush is suitable for use with IPA, trichloroethane and other cleaning solvents.