Learn More
Waters Corp WATERS CORPORATION Cap and Septum, Black, PTFE, 100/pk
Catalog No. NC1476996 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cap and PTFE septum is designed for normal phase solvents, protecting samples from contaminants during sample preparation. This 100-pack of black vial caps and PTFE septa is strictly QC tested for cleanliness properties and dimensional tolerances to ensure proper application.
  • Designed for use with normal phase solvents.
  • Strictly QC tested for cleanliness properties and dimensional tolerances.
Catalog No. NC1476996 Supplier Waters Corp Supplier No. 186007198
This cap and PTFE septum is designed for normal phase solvents, protecting samples from contaminants during sample preparation. This 100-pack of black vial caps and PTFE septa is strictly QC tested for cleanliness properties and dimensional tolerances to ensure proper application.
  • Designed for use with normal phase solvents.
  • Strictly QC tested for cleanliness properties and dimensional tolerances.