Learn More
MSC 80/20 Inc. Button Head Socket Cap Screw, Steel, M8x1.25, 25 mm, 1 ea
Catalog No. 5000512101
Encompass
This button head socket cap screw is a robust fastening component designed for versatile use with 15/30/40 series systems. Constructed from steel with a black zinc-plated finish, it provides reliable structural support in various assembly applications.
  • Features an M8 x 1.25 thread size for precise compatibility
  • Includes a full thread configuration for secure fastening
  • Built from durable steel material with a protective finish
  • Designed for integration with 15, 30, and 40 series profiles
  • ESD safe for use in sensitive environments
  • Overall length of 25.00 mm
Catalog No. 50-005-12101 Supplier MSC Supplier No. 54976659
This button head socket cap screw is a robust fastening component designed for versatile use with 15/30/40 series systems. Constructed from steel with a black zinc-plated finish, it provides reliable structural support in various assembly applications.
  • Features an M8 x 1.25 thread size for precise compatibility
  • Includes a full thread configuration for secure fastening
  • Built from durable steel material with a protective finish
  • Designed for integration with 15, 30, and 40 series profiles
  • ESD safe for use in sensitive environments
  • Overall length of 25.00 mm