Learn More
World Precision Instrument Black Coated Westcott Scissors
SDP
Supplier:  World Precision Instrument WPB4041
Encompass
Black-Coated Westcott Spring Scissors are popular spring handled scissors that automatically reopen after each cut. Westcott brand spring scissors give you an ergonomic and comfortable grip, reducing hand strain and fatigue during repetitive cutting tasks. Westcott spring scissors are perfect for ambidextrous use, often preferred by left-handed individuals. The non-reflective black coating makes these more comfortable to use under a microscope or bright surgical lights. The black coating is also resilient and durable, making for stronger blades and a pair of scissors that last much longer. They are available with blunt, curved or sharp, curved tips. These are constructed of surgical grade stainless steel and are suitable for autoclaving or chemical sterilization.
Catalog No. 50-253-8271