Learn More
Sklar Instruments Sklar Black DeBakey Forceps, Sklar, 9-1/2 in., 2mm, Straight, 1X2 Rows of Nontraumatic Fine Teeth, Stainless Steel with Reduced Reflectivity Polymer Coating. EA/1.
SDP
Catalog No. 501436570 Shop All Sklar Instruments Products
Encompass_Preferred
Sklar Black DeBakey Forceps, Sklar, 9-1/2 in., 2mm, Straight, 1X2 Rows of Nontraumatic Fine Teeth, Stainless Steel with Reduced Reflectivity Polymer Coating. Sklar Black DeBakey Atraumatic Tissue Forceps are a versatile thumb forceps used for grasping fine tissue during laser surgeries. It is available in multiple lengths, straight or angled, and with a variety of tip dimensions. They are commonly used in vascular and general surgery. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements.
This item is not returnable. View return policy
Catalog No. 50-143-6570 Supplier Sklar Instruments Supplier No. 902312
Sklar Black DeBakey Forceps, Sklar, 9-1/2 in., 2mm, Straight, 1X2 Rows of Nontraumatic Fine Teeth, Stainless Steel with Reduced Reflectivity Polymer Coating. Sklar Black DeBakey Atraumatic Tissue Forceps are a versatile thumb forceps used for grasping fine tissue during laser surgeries. It is available in multiple lengths, straight or angled, and with a variety of tip dimensions. They are commonly used in vascular and general surgery. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements.