Learn More
MSC Perma-Patch Asphalt Patch, Black, 5 gal, Zero VOC
Supplier:  MSC 53354437
Encompass
Perma-Patch Black Fill is an all-in-one asphalt patch combining crack filling with surface sealing for pavement repair. This asphaltic emulsion blend contains mineral fillers, recycled shingles, plasticizers, and HDPE fibers, filling wide cracks and repairing alligatored areas. It provides a durable, sealed wear surface in a single application, suitable for various asphaltic surfaces, and resists automotive chemicals. Ready for foot traffic in one hour and vehicular traffic in two hours under optimal conditions.
  • Fills cracks and seals surfaces.
  • Asphaltic emulsion with mineral fillers, recycled shingles, and fibers.
  • Durable, sealed wear surface in one application.
  • Ideal for driveways, parking lots, highways.
  • Excellent color retention; resists automotive chemicals.
  • Foot traffic ready in 1 hour, vehicular in 2 hours.
  • Zero VOCs, PAH compliant, no coal tar.
Catalog No. 50-283-8358