Learn More
MSC Perma-Patch Black Fill, Asphalt Emulsion, 55 Gallon Drum, Black
Catalog No. 502837489
Encompass
This asphaltic emulsion blend is an all-in-one solution for asphalt alligatoring, combining crack filling and surface sealing. Formulated with mineral fillers, recycled shingles, plasticizers, and HDPE fibers, it repairs alligatored areas and provides a durable wear surface.
  • Effortless application via roller or spray.
  • Rapid ready times: walk on in one hour, drive on in two hours.
  • Year-round performance, effective from 40°F to 300°F.
  • Remains flexible in cold and resists softening in heat.
  • Non-skid, jet-black surface with one-coat protection.
  • Eco-friendly: zero VOCs, no coal tar, and PAH compliant.
  • Resistant to de-icing salts and automotive chemicals.
  • Ready-to-use; simply stir and pour.
Catalog No. 50-283-7489 Supplier MSC Supplier No. 72194145
This asphaltic emulsion blend is an all-in-one solution for asphalt alligatoring, combining crack filling and surface sealing. Formulated with mineral fillers, recycled shingles, plasticizers, and HDPE fibers, it repairs alligatored areas and provides a durable wear surface.
  • Effortless application via roller or spray.
  • Rapid ready times: walk on in one hour, drive on in two hours.
  • Year-round performance, effective from 40°F to 300°F.
  • Remains flexible in cold and resists softening in heat.
  • Non-skid, jet-black surface with one-coat protection.
  • Eco-friendly: zero VOCs, no coal tar, and PAH compliant.
  • Resistant to de-icing salts and automotive chemicals.
  • Ready-to-use; simply stir and pour.