Learn More
MSC Vermont Gage Black Guard Pin Gauge, 0.1380 in, Class ZZ, Minus, 1 ea
Catalog No. 5000512160
Encompass
This Black Guard™ cylindrical pin gage is designed for high-precision measurement of hole sizes. It is a Class ZZ gage featuring a minus tolerance, ensuring accurate go/no-go gauging in metalworking and inspection applications. The gage is constructed from durable tool steel with a 60/65 Rc hardness rating and a surface finish of 10 microinch or better.
  • Nominal diameter of 0.1380 in.
  • Tolerance of +0.0000 in. to -0.0002 in.
  • Overall length of 2 in.
  • Includes certificate of accuracy with NIST traceability.
  • Reversible design for extended service life.
  • Marked with size and direction of tolerance.
  • Compliant with ISO 17025 standards.
Catalog No. 50-005-12160 Supplier MSC Supplier No. 62257381
This Black Guard™ cylindrical pin gage is designed for high-precision measurement of hole sizes. It is a Class ZZ gage featuring a minus tolerance, ensuring accurate go/no-go gauging in metalworking and inspection applications. The gage is constructed from durable tool steel with a 60/65 Rc hardness rating and a surface finish of 10 microinch or better.
  • Nominal diameter of 0.1380 in.
  • Tolerance of +0.0000 in. to -0.0002 in.
  • Overall length of 2 in.
  • Includes certificate of accuracy with NIST traceability.
  • Reversible design for extended service life.
  • Marked with size and direction of tolerance.
  • Compliant with ISO 17025 standards.