Learn More
Beckman Coulter Beckman Coulter Black Hytrel Tubing, 1/2 in ID, 1 ft - Replacement for MET ONE 3400 Series
Catalog No. NC3454564
Encompass
Beckman Coulter black Hytrel tubing (MPN 480-100-0041) is a flexible, chemically resistant replacement tubing sold per foot for use with MET ONE 3400 series particle counters and similar low-flow fluid or air-handling systems.
  • Flexible Hytrel thermoplastic polyester elastomer
  • Black color
  • 1/2 in inner diameter
  • Sold per foot
  • Chemical and abrasion resistant
  • Suitable for low-flow particle counter and fluid-handling applications
Catalog No. NC3454564 Supplier Beckman Coulter Supplier No. 4801000041
Beckman Coulter black Hytrel tubing (MPN 480-100-0041) is a flexible, chemically resistant replacement tubing sold per foot for use with MET ONE 3400 series particle counters and similar low-flow fluid or air-handling systems.
  • Flexible Hytrel thermoplastic polyester elastomer
  • Black color
  • 1/2 in inner diameter
  • Sold per foot
  • Chemical and abrasion resistant
  • Suitable for low-flow particle counter and fluid-handling applications