Learn More
Alkali Scientific Black Immunoplates PS 96 Well
SDP
Catalog No. 503028790
Encompass_Preferred
ASI provides high quality 96-well plates suitable for various high throughput assays Fluorescence and luminescence assays have been successfully tested with ASI 96 Well Microplates by many high throughput laboratories SPL 96-well microplates have a characteristic feature of high signal with low background White plates provide maximum reflection and minimum crosstalk for luminescence assay Black plates are designed for minimum light scattering and crosstalk during fluorescence assay Maxibinding Modified polystyrene surface for higher binding capacity for proteins and other molecules with both hydrophilic and hydrophobic regions Unibinding Suitable for adsorption of hydrophobic molecules Suitable for fluorescence assay luminescence assay 2 type surfaces Individual lids provided for each plate (Black Plate with black lid White plate with transparent lid) Plate type (96well per plate) HTS (High-Throughput Screening) compatible Flat
Catalog No. 50-302-8790 Supplier Alkali Scientific Supplier No. TMB2096
Add to Cart
Add to Cart
ASI provides high quality 96-well plates suitable for various high throughput assays Fluorescence and luminescence assays have been successfully tested with ASI 96 Well Microplates by many high throughput laboratories SPL 96-well microplates have a characteristic feature of high signal with low background White plates provide maximum reflection and minimum crosstalk for luminescence assay Black plates are designed for minimum light scattering and crosstalk during fluorescence assay Maxibinding Modified polystyrene surface for higher binding capacity for proteins and other molecules with both hydrophilic and hydrophobic regions Unibinding Suitable for adsorption of hydrophobic molecules Suitable for fluorescence assay luminescence assay 2 type surfaces Individual lids provided for each plate (Black Plate with black lid White plate with transparent lid) Plate type (96well per plate) HTS (High-Throughput Screening) compatible Flat