Learn More
Ted Pella Inc Black Nail Polish
SDP
Catalog No. NC0381432
Encompass
Black Nail Polish is used to attach and seal troughs and boats or tape boats for sectioning. The nonreflective background provides easy visibility, and the stickiness provides complete sealing and no interference in sectioning after it dries. Black Nail Polish also offers the ability to secure eyelashes or fine hair to wooden sticks for section manipulation.
Catalog No. NC0381432 Supplier Ted Pella Inc Supplier No. 1148
Add to Cart
Add to Cart
Black Nail Polish is used to attach and seal troughs and boats or tape boats for sectioning. The nonreflective background provides easy visibility, and the stickiness provides complete sealing and no interference in sectioning after it dries. Black Nail Polish also offers the ability to secure eyelashes or fine hair to wooden sticks for section manipulation.