Learn More
Analytical Sales & Services Black Open Top PP Cap with PTFE/Silicone (0.065”) Setpa, 15-425mm Thread
Catalog No. 5000804663
Encompass
Black Open Top Polypropylene Cap with PTFE/Silicone (0.065”) Setpa, 15-425mm Thread
Catalog No. 50-008-04663 Supplier Analytical Sales & Services Supplier No. 31543CASE
Black Open Top Polypropylene Cap with PTFE/Silicone (0.065”) Setpa, 15-425mm Thread