Learn More
Description
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Amber Glass Bottle |
| Refractive Index | 1.479-1.488 |
| Quantity | 500 mL |
| UN Number | UN1197 |
| Specific Gravity | 0.864-0.884 |
| Grade | FCC |
| Physical Form | Viscous Liquid or Low Melting Solid |
| Chemical Name or Material | Black Pepper Oil |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
