Learn More
Fisherbrand™ Black Phenolic PolyCone Cap
Catalog No. 02912062 Shop All Fisherbrand Products
Change view
Click to view available options
Neck Finish:
33-400
Quantity:
576/Cs.
Catalog No. Neck Finish Quantity
02-912-062 33-400 576/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 02-912-062
This item is not returnable. View return policy
Forms an exceptionally tight seal and offer a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.
PolyCone Lined Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier. This liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off. Great for everything from liquids, to soaps, to macro-invertebrates.
  • Liner molds itself around the sealing areas of finish as cap is screwed on
  • Eliminates leakage, evaporation, contamination, binding and backing off
Specifications
Color Black
Material Phenolic Resin
Quantity 576/Cs.
Liner Material PolyCone
Neck Finish 33-400
Product Type Polycone Cap