Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black Phenolic PolyCone Lined Caps

Catalog No. 02993433
Change view
Click to view available options
For Use With (Equipment):
Bottle
Narrow mouth bottle
Wide mouth bottle
Neck Finish:
13-425
15-425
18-400
20-400
22-400
24-400
24-410
28-400
33-400
38-400
No. per Case:
10000 Cs.
1600 Cs.
2200 Cs.
3100 Cs.
5000 Cs.
5500 Cs.
576 Cs.
6500 Cs.
Packaging:
Bagged
Bulk
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Neck Finish No. per Case Packaging
02-993-433 Wide mouth bottle 22-400 576 Cs. Bagged
02-993-438 Narrow mouth bottle 13-425 576 Cs. Bagged
02-993-437 Bottle 24-410 576 Cs. Bagged
02-991-076 Wide mouth bottle 22-400 5000 Cs. Bulk
02-883-63 Wide mouth bottle 18-400 5500 Cs. Bulk
02-993-439 Wide mouth bottle 24-400 576 Cs. Bagged
02-993-440 Wide mouth bottle 38-400 576 Cs. Bagged
36-101-2194 Wide mouth bottle 33-400 576 Cs. Bagged
02-993-436 Wide mouth bottle 18-400 576 Cs. Bagged
02-993-434 Narrow mouth bottle 15-425 576 Cs. Bagged
02-991-174 Wide mouth bottle 28-400 3100 Cs. Bulk
02-991-074 Wide mouth bottle 18-400 6500 Cs. Bulk
02-993-435 Wide mouth bottle 20-400 576 Cs. Bagged
02-991-097 Wide mouth bottle 33-400 2200 Cs. Bulk
02-993-441 Wide mouth bottle 28-400 576 Cs. Bagged
02-991-133 Narrow mouth bottle 15-425 10000 Cs. Bulk
02-991-078 Wide mouth bottle 38-400 1600 Cs. Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 02-993-433 Supplier Qorpak™ Supplier No. CAP00161
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Forms an exceptionally tight seal and offer a good chemical barrier. Qorpak™ Black Phenolic PolyCone Lined Caps is used for everything from liquids, to soaps, to macro-invertebrates.

  • Molded liner around the sealing areas eliminates leakage, evaporation, contamination, binding and backing off
  • Available in ribbed style
  • Available in bulk/bagged packaging

Specifications

Color Black
Material Phenolic Resin
No. per Case 576 Cs.
No. per Pack 12/Pk.
Packaging Bagged
Sterility Non-sterile
Liner Material Polycone
Neck Finish 22-400
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Lined Cap (Ribbed)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.