Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black Phenolic Pulp/Aluminum Foil Lined Caps

Catalog No. 02991256
Change view
Click to view available options
Neck Finish:
13-425
22-400
24-400
33-400
38-400
48-400
58-400
Product Type:
Lined Cap (Ribbed)
Lined Cap (Smooth)
No. per Case:
1248 Cs.
13500 Cs.
288 Cs.
4600 Cs.
576 Cs.
Packaging:
Bagged
Bulk
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Product Type No. per Case Packaging
02-991-256 13-425 Lined Cap (Ribbed) 13500 Cs. Bulk
02-991-135 48-400 Lined Cap (Smooth) 1248 Cs. Bulk
02-993-230 58-400 Lined Cap (Smooth) 288 Cs. Bagged
02-991-431 22-400 Lined Cap (Smooth) 4600 Cs. Bulk
02-993-249 13-425 Lined Cap (Ribbed) 576 Cs. Bagged
02-993-227 38-400 Lined Cap (Smooth) 576 Cs. Bagged
02-993-239 48-400 Lined Cap (Smooth) 576 Cs. Bagged
02-993-228 33-400 Lined Cap (Smooth) 576 Cs. Bagged
02-993-245 24-400 Lined Cap (Smooth) 576 Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 02-991-256 Supplier Qorpak™ Supplier No. CAP00106
Add to Cart
Add to Cart

Ideal for non-acid and non-alkaline products. Qorpak™ Black Phenolic Pulp/Aluminum Foil Lined Caps are typically used for food applications.

  • Closure liners free of odor and taste
  • Used for organic solvents, chrome cleaners, brake fluids and mineral oils
  • Neck finish for narrow bottles is 425 GPI and 400 for wider
  • Available as ribbed style
  • Available in bulk/bagged packaging

Specifications

Color Black
Material Phenolic Resin
No. per Case 13500 Cs.
Packaging Bulk
Sterility Non-sterile
Liner Material Pulp/Aluminum Foil
Neck Finish 13-425
Disposable Reusable
Product Type Lined Cap (Ribbed)
For Use With (Equipment) Narrow mouth bottle
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.