Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black Phenolic Rubber Lined Caps

Catalog No. 02991265
Change view
Click to view available options
Neck Finish:
13-425
15-425
22-400
24-414
28-400
43-400
Product Type:
Lined Cap (Ribbed)
Lined Cap (Smooth)
No. per Case:
11000 Cs.
13500 Cs.
3000 Cs.
576 Cs.
Packaging:
Bagged
Bulk
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Product Type No. per Case Packaging
02-991-265 15-425 Lined Cap (Ribbed) 11000 Cs. Bulk
02-991-342 13-425 Lined Cap (Ribbed) 13500 Cs. Bulk
02-993-356 43-400 Lined Cap (Smooth) 576 Cs. Bagged
02-991-099 28-400 Lined Cap (Smooth) 3000 Cs. Bulk
02-993-351 24-414 Lined Cap (Ribbed) 576 Cs. Bagged
02-993-299 22-400 Lined Cap (Smooth) 576 Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 02-991-265 Supplier Qorpak™ Supplier No. CAP00470
Add to Cart
Add to Cart
Request a Quote

Intented for repeated use. Qorpak™ Black Phenolic Rubber Lined Caps are autoclavable.

  • Liner exhibit resiliency
  • Resistant to moisture vapor
  • Provides consistent adherence to close dimensional tolerance
  • Available in ribbed and smooth style
  • Available as bulk/bagged packaging

Compliance

FDA 21 CFR177.2600

Specifications

Color Black
Material Phenolic Resin
No. per Case 11000 Cs.
Packaging Bulk
Sterility Non-sterile
Liner Material Rubber
Neck Finish 15-425
Certifications/Compliance FDA 21 CFR 177.2600
Disposable Reusable
Product Type Lined Cap (Ribbed)
For Use With (Equipment) Narrow mouth bottle
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.