Learn More
Electron Microscopy Sciences Black Picking Tray, ABS Plastic
SDP
Supplier:  Electron Microscopy Sciences 7117101
Encompass_Preferred
Trays perfect for placement of grids and storage of small components. The trays feature 42 cells with a lip differentiating each cell. The cells measure 11 mm x 11 mm (7/16 x 7/16").
Material: ABS Plastic
Catalog No. 50-193-0109