Learn More
Grainger Black Pipe Nipple Threaded 1/2x1-1/8 In
Catalog No. 502749528
Encompass_Preferred
The Beck Nipple is a black anodized steel nipple fitting with National Pipe Thread (NPT) connections. It has a standard weight wall and is made from ASTM A53 carbon steel. This nipple has 1/2 inch NPT straight thread connections and is 2 inches in length. It can be used to connect and extend threaded pipe. The maximum pressure rating is 6000 psi at 100°F. This nipple fitting is manufactured in the USA by Beck Manufacturing in accordance with ANSI standards. It weighs approximately 0.22 lbs.
Catalog No. 50-274-9528 Supplier Grainger Supplier No. 14J777
The Beck Nipple is a black anodized steel nipple fitting with National Pipe Thread (NPT) connections. It has a standard weight wall and is made from ASTM A53 carbon steel. This nipple has 1/2 inch NPT straight thread connections and is 2 inches in length. It can be used to connect and extend threaded pipe. The maximum pressure rating is 6000 psi at 100°F. This nipple fitting is manufactured in the USA by Beck Manufacturing in accordance with ANSI standards. It weighs approximately 0.22 lbs.