Learn More
Grainger Black Pipe Nipple Threaded 1/2x1-1/2 In
Catalog No. 502750671
Encompass_Preferred
The Beck Nipple is a black anodized steel nipple with National Pipe Thread (NPT) connections. It has a pressure rating of 2000 psi at 70°F and is made from ASTM A733 carbon steel. The nipple has a wall thickness of 0.154 inches, is 3 inches in length, and has 1/2 inch NPT threads on both ends. It can be used to connect and extend threaded pipe. The nipple is black anodized for corrosion resistance.
Catalog No. 50-275-0671 Supplier Grainger Supplier No. 14J778
The Beck Nipple is a black anodized steel nipple with National Pipe Thread (NPT) connections. It has a pressure rating of 2000 psi at 70°F and is made from ASTM A733 carbon steel. The nipple has a wall thickness of 0.154 inches, is 3 inches in length, and has 1/2 inch NPT threads on both ends. It can be used to connect and extend threaded pipe. The nipple is black anodized for corrosion resistance.