Learn More
Grainger Beck Pipe Nipple, Seamless, Schedule XXH, 1/2 in., 2-1/2 in. Long
Catalog No. 502750956
Encompass_Preferred
This black steel pipe nipple is a seamless, heavy-duty component designed for robust industrial and commercial applications. Manufactured from ASTM A106 Grade B material and conforming to Schedule XXH specifications, this nipple is engineered for high-pressure systems transporting air, gases, non-potable water, oils, and steam. It features male NPT threaded connections on both ends for secure and reliable joint assembly.
  • Seamless construction meeting ASTM A733 standards
  • Nominal pipe size of 1/2 in. with an overall length of 2-1/2 in.
  • Compatible with Schedule XXH pipe systems
  • Constructed from high-grade black steel
  • Standardized NPT threads per ASME B1.20.1 for precise compatibility
Catalog No. 50-275-0956 Supplier Grainger Supplier No. 14J780
This black steel pipe nipple is a seamless, heavy-duty component designed for robust industrial and commercial applications. Manufactured from ASTM A106 Grade B material and conforming to Schedule XXH specifications, this nipple is engineered for high-pressure systems transporting air, gases, non-potable water, oils, and steam. It features male NPT threaded connections on both ends for secure and reliable joint assembly.
  • Seamless construction meeting ASTM A733 standards
  • Nominal pipe size of 1/2 in. with an overall length of 2-1/2 in.
  • Compatible with Schedule XXH pipe systems
  • Constructed from high-grade black steel
  • Standardized NPT threads per ASME B1.20.1 for precise compatibility