Learn More
Grainger Beck Black Steel Pipe Nipple, 1/2 in, Schedule XXH, Seamless, 1 ea
Catalog No. 502750939
Encompass_Preferred
This 1/2 in. nominal pipe size black carbon steel nipple is designed for industrial applications including the transport of air, gas, non-potable water, oil, and steam. It features a seamless construction for enhanced durability and is threaded on both ends with male NPT connections.
  • Manufactured from carbon steel confirming to ASTM A106 Grade B
  • Conforms to ASTM A733 standards for seamless steel nipples
  • NPT threads adhere to ASME B1.20.1
  • Black finish
  • 2 in. overall length
  • Suitable for air, gas, non-potable water, oil, and steam applications
Catalog No. 50-275-0939 Supplier Grainger Supplier No. 14J779
This 1/2 in. nominal pipe size black carbon steel nipple is designed for industrial applications including the transport of air, gas, non-potable water, oil, and steam. It features a seamless construction for enhanced durability and is threaded on both ends with male NPT connections.
  • Manufactured from carbon steel confirming to ASTM A106 Grade B
  • Conforms to ASTM A733 standards for seamless steel nipples
  • NPT threads adhere to ASME B1.20.1
  • Black finish
  • 2 in. overall length
  • Suitable for air, gas, non-potable water, oil, and steam applications