Learn More
Grainger Black Pipe Nipple Threaded 1x1-1/2 In
Catalog No. 502752692
Encompass_Preferred
The 1-1/2" Black Iron Pipe Nipple from Grainger is a threaded pipe fitting made from black iron. It has National Pipe Thread ends and is 1-1/2" in diameter. The nipple has a standard wall thickness and is 12" in length. It can be used to extend and connect black iron piping in gas, steam, air or water applications. The working pressure is 850 PSI at 70°F. It meets ASME B16.3 specifications. The nipple is made in the USA.
Catalog No. 50-275-2692 Supplier Grainger Supplier No. 14J803
The 1-1/2" Black Iron Pipe Nipple from Grainger is a threaded pipe fitting made from black iron. It has National Pipe Thread ends and is 1-1/2" in diameter. The nipple has a standard wall thickness and is 12" in length. It can be used to extend and connect black iron piping in gas, steam, air or water applications. The working pressure is 850 PSI at 70°F. It meets ASME B16.3 specifications. The nipple is made in the USA.