Learn More
Grainger Black Pipe Nipple Threaded 1x10 In
Catalog No. 502752606
Encompass_Preferred
The Grainger 14J811 is a 14-inch diameter, 8-inch tall, plastic funnel made in the USA. It has a wide mouth opening for quick pouring and is constructed of durable HDPE plastic. The funnel has a ribbed exterior to prevent slipping and splashing. It can withstand temperatures from -20°F to 200°F. The funnel has a max capacity of 1 gallon and weighs 0.19 lbs. It is ideal for automotive, industrial, and food applications. The funnel is yellow in color.
Catalog No. 50-275-2606 Supplier Grainger Supplier No. 14J811
The Grainger 14J811 is a 14-inch diameter, 8-inch tall, plastic funnel made in the USA. It has a wide mouth opening for quick pouring and is constructed of durable HDPE plastic. The funnel has a ribbed exterior to prevent slipping and splashing. It can withstand temperatures from -20°F to 200°F. The funnel has a max capacity of 1 gallon and weighs 0.19 lbs. It is ideal for automotive, industrial, and food applications. The funnel is yellow in color.