Learn More
Grainger Beck Black Pipe Nipple, Steel, 1 in. Nominal, Schedule XXH
Catalog No. 502751624
Encompass_Preferred
The black steel pipe nipple is a durable, seamless fitting designed for secure connections in industrial piping systems. This component is constructed to withstand demanding environments, making it suitable for a variety of applications involving air, gases, non-potable water, oils, and steam.
  • Constructed from durable black steel
  • Seamless design for enhanced integrity
  • Threaded on both ends with male NPT connections
  • Designed to meet ASTM A106 Grade B and ASTM A733 standards
  • Suitable for air, gas, oil, steam, and non-potable water applications
Catalog No. 50-275-1624 Supplier Grainger Supplier No. 14J805
The black steel pipe nipple is a durable, seamless fitting designed for secure connections in industrial piping systems. This component is constructed to withstand demanding environments, making it suitable for a variety of applications involving air, gases, non-potable water, oils, and steam.
  • Constructed from durable black steel
  • Seamless design for enhanced integrity
  • Threaded on both ends with male NPT connections
  • Designed to meet ASTM A106 Grade B and ASTM A733 standards
  • Suitable for air, gas, oil, steam, and non-potable water applications