Learn More
Grainger Black Pipe Nipple Threaded 1x5 In
Catalog No. 502751338
Encompass_Preferred
The Grainger 14J808 is a 14-inch diameter, 8-inch deep plastic pail with a bail handle and a tight-fitting lid. It has a capacity of 5 gallons and is made from high density polyethylene in natural white color. The pail has graduations on the side for measuring contents. It can be used for a variety of applications including mixing, storing, and transporting viscous materials, liquids, powders, pellets, and small parts. The pail is chemical and corrosion resistant. It meets FDA requirements for food contact.
Catalog No. 50-275-1338 Supplier Grainger Supplier No. 14J808
The Grainger 14J808 is a 14-inch diameter, 8-inch deep plastic pail with a bail handle and a tight-fitting lid. It has a capacity of 5 gallons and is made from high density polyethylene in natural white color. The pail has graduations on the side for measuring contents. It can be used for a variety of applications including mixing, storing, and transporting viscous materials, liquids, powders, pellets, and small parts. The pail is chemical and corrosion resistant. It meets FDA requirements for food contact.