Learn More
Grainger Black Pipe Nipple Threaded 2x2-1/2 In
Catalog No. 502752604
Encompass_Preferred
The Black Pipe Nipple Threaded 2x2 1/2 In is a 2 inch long black steel pipe nipple with National Pipe Thread (NPT) on both ends. It has a 2 inch nominal pipe size (NPS) and is made from ASTM A53 Grade B ERW steel. This nipple allows you to connect and extend black steel pipes. It is zinc coated for corrosion resistance. The working pressure is 850 psi and temperature range is -20 to 450°F. It meets ASME B16.3 specifications. This threaded nipple provides a strong and durable threaded pipe connection.
Catalog No. 50-275-2604 Supplier Grainger Supplier No. 14J818
The Black Pipe Nipple Threaded 2x2 1/2 In is a 2 inch long black steel pipe nipple with National Pipe Thread (NPT) on both ends. It has a 2 inch nominal pipe size (NPS) and is made from ASTM A53 Grade B ERW steel. This nipple allows you to connect and extend black steel pipes. It is zinc coated for corrosion resistance. The working pressure is 850 psi and temperature range is -20 to 450°F. It meets ASME B16.3 specifications. This threaded nipple provides a strong and durable threaded pipe connection.