Learn More
Grainger Black Pipe Nipple Threaded 3/4x10 In
Catalog No. 502750399
Encompass_Preferred
The Grainger Approved Black Pipe Nipple is made from carbon steel and has a black finish. It is 1/2 inch in diameter and 6 inches long. The nipple has National Pipe Thread ends that allow it to connect to other NPT threaded fittings and pipes. It can be used in plumbing, machinery, and hydraulic applications that require a short section of pipe to join fittings. The working pressure is 850 PSI at 70°F. It meets ASME B16.3 specifications. The nipple is made in the USA.
Catalog No. 50-275-0399 Supplier Grainger Supplier No. 14J799
The Grainger Approved Black Pipe Nipple is made from carbon steel and has a black finish. It is 1/2 inch in diameter and 6 inches long. The nipple has National Pipe Thread ends that allow it to connect to other NPT threaded fittings and pipes. It can be used in plumbing, machinery, and hydraulic applications that require a short section of pipe to join fittings. The working pressure is 850 PSI at 70°F. It meets ASME B16.3 specifications. The nipple is made in the USA.