Learn More
Grainger Black Pipe Nipple Threaded 3/4x12 In
Catalog No. 502750392
Encompass_Preferred
The Grainger Approved 1/2 in. Black Iron Pipe Nipple is made from seamless carbon steel pipe. It has National Pipe Thread ends that comply with ANSI standards. This nipple has a pressure rating of 850 psi at 70°F and is for use in general purpose low pressure applications. It can be used to extend and connect black iron pipe. The nipple measures 6 in. long with an inside diameter of 1/2 in. It is zinc coated for corrosion resistance. This product is made in the USA.
Catalog No. 50-275-0392 Supplier Grainger Supplier No. 14J802
The Grainger Approved 1/2 in. Black Iron Pipe Nipple is made from seamless carbon steel pipe. It has National Pipe Thread ends that comply with ANSI standards. This nipple has a pressure rating of 850 psi at 70°F and is for use in general purpose low pressure applications. It can be used to extend and connect black iron pipe. The nipple measures 6 in. long with an inside diameter of 1/2 in. It is zinc coated for corrosion resistance. This product is made in the USA.