Learn More
Grainger Black Pipe Nipple Threaded 3/4x5 In
Catalog No. 502751459
Encompass_Preferred
The Grainger Approved 1/2 in. Black Iron Pipe Nipple is made from malleable iron and has National Pipe Thread ends. It is 6 in. long and can be used to extend and connect black iron piping. This nipple meets ASME B16.3 specifications and is rated to 300 psi WOG at ambient temperatures. It features a black finish to resist rust and corrosion. The nipple's iron construction provides durability while its short length allows for convenient installations. This product enables users to make secure threaded black iron pipe connections.
Catalog No. 50-275-1459 Supplier Grainger Supplier No. 14J794
The Grainger Approved 1/2 in. Black Iron Pipe Nipple is made from malleable iron and has National Pipe Thread ends. It is 6 in. long and can be used to extend and connect black iron piping. This nipple meets ASME B16.3 specifications and is rated to 300 psi WOG at ambient temperatures. It features a black finish to resist rust and corrosion. The nipple's iron construction provides durability while its short length allows for convenient installations. This product enables users to make secure threaded black iron pipe connections.