Learn More
Grainger Black Pipe Nipple Threaded 3/4x7 In
Catalog No. 502751629
Encompass_Preferred
The 3/4" x 7" Black Pipe Nipple is a threaded pipe fitting made from black steel. It has National Pipe Thread ends and is 7 inches long. The nipple has a pressure rating of 2000 psi at 70°F and is commonly used in plumbing, irrigation, and industrial piping applications. The nipple is made in the USA and conforms to ASTM A733 standards. It can join other threaded fittings and valves in piping systems that convey water, air, natural gas, or other fluids.
Catalog No. 50-275-1629 Supplier Grainger Supplier No. 14J796
The 3/4" x 7" Black Pipe Nipple is a threaded pipe fitting made from black steel. It has National Pipe Thread ends and is 7 inches long. The nipple has a pressure rating of 2000 psi at 70°F and is commonly used in plumbing, irrigation, and industrial piping applications. The nipple is made in the USA and conforms to ASTM A733 standards. It can join other threaded fittings and valves in piping systems that convey water, air, natural gas, or other fluids.