Learn More
Grainger Black Pipe Nipple Threaded 3/4x9 In
Catalog No. 502751630
Encompass_Preferred
The 3/4" x 9" Black Pipe Nipple is a threaded pipe fitting made from black malleable iron. It has National Pipe Thread ends and is 9 inches long. This nipple allows you to extend pipe runs and make connections. It is rated for 300 psi working pressure at up to 250°F. The nipple is black in color and weighs approximately 0.75 lbs. This product can be used in steam, water, air, and oil applications. It meets ASTM A197 specifications.
Catalog No. 50-275-1630 Supplier Grainger Supplier No. 14J798
The 3/4" x 9" Black Pipe Nipple is a threaded pipe fitting made from black malleable iron. It has National Pipe Thread ends and is 9 inches long. This nipple allows you to extend pipe runs and make connections. It is rated for 300 psi working pressure at up to 250°F. The nipple is black in color and weighs approximately 0.75 lbs. This product can be used in steam, water, air, and oil applications. It meets ASTM A197 specifications.