Learn More
Grainger Black Pipe Nipple Threaded 3/4x11 In
Catalog No. 502751631
Encompass_Preferred
The Grainger Approved 1/2 in. Black Iron Pipe Nipple is made from seamless carbon steel pipe. It has National Pipe Thread ends that comply with ANSI standards. The nipple has a pressure rating of 850 psi at 70°F. It can be used in steam, air, gas, oil, and water applications. The nipple's working temperature range is -20° to 800°F. It measures 6 in. long and has an outside diameter of 1/2 in. The nipple weighs 0.22 lbs.
Catalog No. 50-275-1631 Supplier Grainger Supplier No. 14J801
The Grainger Approved 1/2 in. Black Iron Pipe Nipple is made from seamless carbon steel pipe. It has National Pipe Thread ends that comply with ANSI standards. The nipple has a pressure rating of 850 psi at 70°F. It can be used in steam, air, gas, oil, and water applications. The nipple's working temperature range is -20° to 800°F. It measures 6 in. long and has an outside diameter of 1/2 in. The nipple weighs 0.22 lbs.