Learn More
Grainger Black Pipe Nipple Threaded 3/4x8 In
Catalog No. 502752810
Encompass_Preferred
The Grainger Approved Black Pipe Nipple is made from carbon steel and has National Pipe Thread ends. It is available in sizes ranging from 1/8 inch to 2 inches diameter, and lengths from 1 inch to 12 inches. The nipple has a working pressure of 850 PSI at 70°F and is compatible with Schedule 40 black pipe and fittings. It can be used in steam, air, gas, or water applications. The nipple is zinc coated for corrosion resistance.
Catalog No. 50-275-2810 Supplier Grainger Supplier No. 14J797
The Grainger Approved Black Pipe Nipple is made from carbon steel and has National Pipe Thread ends. It is available in sizes ranging from 1/8 inch to 2 inches diameter, and lengths from 1 inch to 12 inches. The nipple has a working pressure of 850 PSI at 70°F and is compatible with Schedule 40 black pipe and fittings. It can be used in steam, air, gas, or water applications. The nipple is zinc coated for corrosion resistance.