Learn More
MilliporeSigma™ Supelco™ Black Plastic Caps with White Rubber Liner
Catalog No. 111011123 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Closure Size:
13-425
15-425
20-400
8-425
No. per Pack:
100/Pk.
1000/Pk.
200/Pk.
500/Pk.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size No. per Pack
11-101-1123 13-425 200/Pk.
11-100-9705 8-425 1000/Pk.
11-101-0958 15-425 200/Pk.
11-101-1972 8-425 100/Pk.
11-100-9009 20-400 500/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11-101-1123 Supplier MilliporeSigma™ Supelco™ Supplier No. Z106402200EA
Add to Cart
Add to Cart
Autoclavable caps designed to ensure the ultimate in product safety
  • Solid top
  • White rubber liner
  • Prevents contamination
  • Easy sterilization
TRUSTED_SUSTAINABILITY
Specifications
Color (Seal) White
Material (Liner) Rubber
Color Black
Material Phenolic
No. per Pack 200/Pk.
Quantity 200 Ea.
Closure Size 13-425
Autoclavable Autoclavable
Product Type Solid Top Cap