Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black Polypropylene Pulp/Vinyl Lined Caps

Catalog No. 02991041
Change view
Click to view available options
Packaging:
Bulk
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
02-991-041 Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-991-041 Supplier Qorpak™ Supplier No. CAP00097
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for food, beverage, medical and chemical applications packed at <120°F. Qorpak™ Black Polypropylene Pulp/Vinyl Lined Caps are ideal for general use.

  • HDPE coated paper liner
  • Used for mild acids, alkalis, solvents, alcohols, oils and aqueous products
  • Not recommended for active hydrocarbons or bleaches
  • Available in smooth and ribbed style
  • Available as bulk/bagged packaging

Specifications

Color Black
Material Polypropylene
No. per Case 256 Cs.
Packaging Bulk
Sterility Non-sterile
Neck Finish 100-400
For Use With (Equipment) Wide mouth bottle
Closure Liner Pulp/Vinyl
Closure Type Smooth
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Threaded Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.