Learn More
Waters Corp Waters Black Cap and Septum, Preslit PTFE/Silicone, 12 x 32 mm
Catalog No. 50179326 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These black caps and preslit PTFE/Silicone septa are designed as spare caps for TruView LC/MS Certified Vials. The septa are engineered to withstand multiple injections, ensuring reliable and consistent results. For convenience, the caps and septa are pre-assembled for easy and efficient laboratory use. To maintain their integrity and performance, it is recommended to store these components in a cool and dry environment.
  • Designed for TruView LC/MS certified vials.
  • Features a preslit PTFE/Silicone septum.
  • Includes color-coded black caps.
  • Withstands multiple injections.
  • Pre-assembled for convenience.
Catalog No. 50-179-326 Supplier Waters Corp Supplier No. 186005827
These black caps and preslit PTFE/Silicone septa are designed as spare caps for TruView LC/MS Certified Vials. The septa are engineered to withstand multiple injections, ensuring reliable and consistent results. For convenience, the caps and septa are pre-assembled for easy and efficient laboratory use. To maintain their integrity and performance, it is recommended to store these components in a cool and dry environment.
  • Designed for TruView LC/MS certified vials.
  • Features a preslit PTFE/Silicone septum.
  • Includes color-coded black caps.
  • Withstands multiple injections.
  • Pre-assembled for convenience.