Learn More
Perkin Elmer US LLC Standard PVC Three-Stop Peristaltic Pump Tubing - 0.76 mm I.D., Black/Black/Black, Pkg. 12
Supplier:  Perkin Elmer US LLC B0506058
Encompass_Preferred
This Three-Stop Peristaltic Pump Tubing is designed for use with the FIAS, FIMS, and the S10 Autosampler. This clear tubing ships as a package of 12.
Catalog No. 50-905-2012